Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MIS18BP1
Entrez Gene ID
55320
Gene Name
MIS18 binding protein 1
Gene Aliases
C14orf106, HSA242977, KNL2, M18BP1
Chromosome Location
Chr.14: 45203190 - 45253402 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005267833.4 XP_005267890.1 1 - 2 311 87

View Full Product Details