The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001130674.2 | NP_001124146.1 | 3 - 4 | 563 | 110 |
NM_001199672.1 | NP_001186601.1 | 4 - 5 | 1378 | 110 | |
GenBank mRNA | AF257659.1 | - | 2 - 3 | 354 | 110 |
AF345637.1 | - | 3 - 4 | 456 | 110 | |
AK027460.1 | - | 3 - 4 | 492 | 110 | |
HM002605.1 | - | 4 - 5 | 1379 | 110 | |
HM002609.1 | - | 3 - 4 | 563 | 110 | |
HM002615.1 | - | 5 - 6 | 1573 | 110 |