The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001130674.2 | NP_001124146.1 | 6 - 7 | 997 | 125 |
NM_001199671.1 | NP_001186600.1 | 7 - 8 | 1812 | 125 | |
NM_001199672.1 | NP_001186601.1 | 7 - 8 | 1812 | 125 | |
NM_001219.4 | NP_001210.1 | 6 - 7 | 997 | 125 | |
NR_074086.1 | 4 - 5 | 571 | 125 | ||
XM_017012659.1 | XP_016868148.1 | 6 - 7 | 1362 | 125 | |
GenBank mRNA | AF013759.1 | - | 6 - 7 | 903 | 125 |
AF257659.1 | - | 5 - 6 | 788 | 125 | |
AF345637.1 | - | 6 - 7 | 890 | 125 | |
AK027460.1 | - | 6 - 7 | 926 | 125 | |
AK056338.1 | - | 7 - 8 | 1700 | 125 | |
AK074862.1 | - | 6 - 7 | 947 | 125 | |
AK312446.1 | - | 6 - 7 | 940 | 125 | |
BC013383.2 | - | 6 - 7 | 896 | 125 | |
BT006826.1 | - | 5 - 6 | 845 | 125 | |
BT020143.1 | - | 5 - 6 | 845 | 125 | |
CR457039.1 | - | 5 - 6 | 845 | 125 | |
CR541835.1 | - | 5 - 6 | 845 | 125 | |
HM002604.1 | - | 7 - 8 | 1813 | 125 | |
HM002605.1 | - | 7 - 8 | 1813 | 125 | |
HM002606.1 | - | 5 - 6 | 739 | 125 | |
HM002610.1 | - | 4 - 5 | 514 | 125 | |
HM002613.1 | - | 5 - 6 | 802 | 125 | |
HM002615.1 | - | 8 - 9 | 2007 | 125 | |
HM002616.1 | - | 4 - 5 | 571 | 125 | |
U67280.1 | - | 6 - 7 | 907 | 125 |