Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- CREM
- Entrez Gene ID
-
1390
- Gene Name
- cAMP responsive element modulator
- Gene Aliases
-
CREM-2, ICER, hCREM-2
- Chromosome Location
-
Chr.10: 35126841 - 35212958 on Build GRCh38
View Full Product Details