Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SYT7
Entrez Gene ID
9066
Gene Name
synaptotagmin 7
Gene Aliases
IPCA-7, IPCA7, PCANAP7, SYT-VII, SYTVII
Chromosome Location
Chr.11: 61513716 - 61588404 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004200.3 NP_004191.2 3 - 4 269 114
XM_011545343.2 XP_011543645.1 3 - 4 255 114
GenBank mRNA AF038535.1 - 3 - 4 262 114
BC125170.1 - 3 - 4 227 114
BE644642.1 - 3 - 4 269 114

View Full Product Details