The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002128.5 | NP_002119.1 | 1 - 2 | 165 | 179 |
GenBank mRNA | AA430980.1 | - | 1 - 2 | 165 | 179 |