The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001261463.1 | NP_001248392.1 | 28 - 29 | 3381 | 113 |
NM_001322945.1 | NP_001309874.1 | 27 - 28 | 3273 | 113 | |
NM_006175.4 | NP_006166.3 | 27 - 28 | 3276 | 113 | |
NM_198060.3 | NP_932326.2 | 28 - 29 | 3381 | 113 | |
XM_005269864.2 | XP_005269921.1 | 27 - 28 | 3252 | 113 | |
XM_005269865.2 | XP_005269922.1 | 27 - 28 | 3249 | 113 | |
XM_005269867.2 | XP_005269924.1 | 28 - 29 | 3357 | 113 | |
XM_006717870.2 | XP_006717933.1 | 27 - 28 | 3252 | 113 | |
XM_011539832.2 | XP_011538134.1 | 28 - 29 | 3357 | 113 | |
GenBank mRNA | AL831980.2 | - | 28 - 29 | 3308 | 113 |
AL832025.2 | - | 28 - 29 | 3308 | 113 | |
AL832330.2 | - | 28 - 29 | 3308 | 113 | |
AL832457.3 | - | 14 - 15 | 1734 | 113 | |
AY081943.2 | - | 27 - 28 | 3253 | 113 | |
AY177620.1 | - | 28 - 29 | 3334 | 113 | |
AY177621.1 | - | 27 - 28 | 3229 | 113 | |
BC126407.1 | - | 28 - 29 | 3217 | 113 | |
BC143751.1 | - | 28 - 29 | 3217 | 113 | |
BX640682.2 | - | 28 - 29 | 3308 | 113 | |
BX640730.1 | - | 28 - 29 | 3465 | 113 | |
BX641052.1 | - | 28 - 29 | 3345 | 113 |