The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001244580.1 | NP_001231509.1 | 63 - 64 | 9976 | 156 |
NM_003496.3 | NP_003487.1 | 62 - 63 | 9889 | 156 | |
GenBank mRNA | AB209489.1 | - | 54 - 55 | 8899 | 156 |
AF076974.1 | - | 62 - 63 | 9889 | 156 | |
AF110377.1 | - | 63 - 64 | 9896 | 156 |