The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001167738.1 | NP_001161210.1 | 12 - 13 | 2425 | 98 |
NM_020443.4 | NP_065176.3 | 14 - 15 | 3756 | 98 | |
XM_006711609.2 | XP_006711672.2 | 12 - 13 | 2391 | 98 | |
XM_006711610.2 | XP_006711673.2 | 11 - 12 | 2367 | 98 | |
XM_006711611.1 | XP_006711674.1 | 12 - 13 | 2507 | 98 | |
XM_011510097.1 | XP_011508399.1 | 14 - 15 | 3435 | 98 | |
XM_011510098.1 | XP_011508400.1 | 13 - 14 | 3411 | 98 | |
XM_011510099.1 | XP_011508401.1 | 13 - 14 | 3411 | 98 | |
XM_011510100.1 | XP_011508402.1 | 13 - 14 | 3264 | 98 | |
XM_011510101.1 | XP_011508403.1 | 12 - 13 | 3240 | 98 | |
XM_011510102.1 | XP_011508404.1 | 12 - 13 | 3240 | 98 | |
XM_017002751.1 | XP_016858240.1 | 13 - 14 | 3264 | 98 | |
XM_017002752.1 | XP_016858241.1 | 11 - 12 | 2367 | 98 | |
XM_017002753.1 | XP_016858242.1 | 13 - 14 | 3411 | 98 | |
GenBank mRNA | AB032977.3 | - | 14 - 15 | 3089 | 98 |
AB033039.1 | - | 8 - 9 | 1268 | 98 | |
AJ488101.1 | - | 14 - 15 | 3287 | 98 | |
AK022631.1 | - | 10 - 11 | 2158 | 98 | |
AY043013.1 | - | 14 - 15 | 3756 | 98 | |
BX648460.1 | - | 12 - 13 | 2392 | 98 |