The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001199397.1 | NP_001186326.1 | 5 - 6 | 886 | 80 |
NM_001199398.1 | NP_001186327.1 | 5 - 6 | 886 | 80 | |
NM_001199399.1 | NP_001186328.1 | 5 - 6 | 886 | 80 | |
NM_001199400.1 | NP_001186329.1 | 4 - 5 | 964 | 80 | |
NM_012224.2 | NP_036356.1 | 4 - 5 | 964 | 80 | |
XM_006714228.1 | XP_006714291.1 | 5 - 6 | 888 | 80 | |
XM_011532003.1 | XP_011530305.1 | 5 - 6 | 888 | 80 | |
XM_011532004.1 | XP_011530306.1 | 5 - 6 | 888 | 80 | |
XM_011532005.2 | XP_011530307.1 | 5 - 6 | 888 | 80 | |
XM_017008250.1 | XP_016863739.1 | 4 - 5 | 963 | 80 | |
XM_017008253.1 | XP_016863742.1 | 5 - 6 | 888 | 80 | |
GenBank mRNA | AB067488.1 | - | 4 - 5 | 800 | 80 |
AK027580.1 | - | 1 - 2 | 40 | 80 | |
AL050385.1 | - | 5 - 6 | 841 | 80 | |
BC015147.1 | - | 4 - 5 | 726 | 80 | |
BC068201.1 | - | 5 - 6 | 846 | 80 | |
BC114491.1 | - | 5 - 6 | 646 | 80 | |
CR933642.1 | - | 5 - 6 | 841 | 80 | |
CR936672.1 | - | 4 - 5 | 921 | 80 |