Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
WASF1
Entrez Gene ID
8936
Gene Name
WAS protein family member 1
Gene Aliases
SCAR1, WAVE, WAVE1
Chromosome Location
Chr.6: 110099817 - 110189874 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001024934.1 NP_001020105.1 3 - 4 872 139
NM_001024935.1 NP_001020106.1 3 - 4 825 139
NM_001024936.1 NP_001020107.1 2 - 3 727 139
NM_003931.2 NP_003922.1 4 - 5 970 139
XM_005267203.3 XP_005267260.1 5 - 6 968 139
XM_005267205.2 XP_005267262.1 3 - 4 434 139
XM_005267206.2 XP_005267263.1 3 - 4 482 139
XM_005267207.2 XP_005267264.1 2 - 3 297 139
XM_011536233.1 XP_011534535.1 3 - 4 582 139
XM_011536234.1 XP_011534536.1 4 - 5 870 139
XM_011536235.1 XP_011534537.1 4 - 5 928 139
XM_017011437.1 XP_016866926.1 5 - 6 990 139
XM_017011438.1 XP_016866927.1 5 - 6 932 139
XM_017011439.1 XP_016866928.1 3 - 4 523 139
XM_017011440.1 XP_016866929.1 2 - 3 400 139
XM_017011441.1 XP_016866930.1 3 - 4 434 139
XM_017011442.1 XP_016866931.1 3 - 4 728 139
XM_017011443.1 XP_016866932.1 2 - 3 330 139
XM_017011444.1 XP_016866933.1 3 - 4 482 139
GenBank mRNA AF134303.1 - 1 - 2 133 139
AK097838.1 - 4 - 5 521 139
BC044591.1 - 3 - 4 443 139
BI461486.1 - 3 - 4 522 139
D87459.1 - 3 - 4 375 139

View Full Product Details