Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DTHD1
Entrez Gene ID
401124
Gene Name
death domain containing 1
Gene Aliases
-
Chromosome Location
Chr.4: 36281593 - 36347511 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001136536.4 NP_001130008.2 3 - 4 931 155
NM_001170700.2 NP_001164171.1 3 - 4 1086 155
XM_006714014.3 XP_006714077.1 4 - 5 1565 155
XM_011513693.2 XP_011511995.1 4 - 5 1566 155
XM_011513694.2 XP_011511996.1 4 - 5 1503 155
XM_011513695.2 XP_011511997.1 4 - 5 1565 155
XM_011513696.2 XP_011511998.1 3 - 4 949 155
XM_017008191.1 XP_016863680.1 4 - 5 1566 155
XM_017008192.1 XP_016863681.1 4 - 5 1568 155
XM_017008193.1 XP_016863682.1 4 - 5 1567 155
GenBank mRNA AK131492.1 - 3 - 4 924 155
AK304357.1 - 3 - 4 917 155
AK316523.1 - 3 - 4 917 155
BC157885.1 - 3 - 4 1086 155

View Full Product Details