The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005736.3 | NP_005727.1 | 8 - 9 | 1031 | 145 |
GenBank mRNA | AK096085.1 | - | 1 - 2 | 1677 | 145 |
AK297362.1 | - | 8 - 9 | 868 | 145 | |
AK297620.1 | - | 7 - 8 | 925 | 145 | |
AK302072.1 | - | 7 - 8 | 920 | 145 | |
AK312506.1 | - | 8 - 9 | 990 | 145 | |
BC000693.2 | - | 8 - 9 | 967 | 145 | |
BC026016.2 | - | 8 - 9 | 933 | 145 | |
X82206.1 | - | 8 - 9 | 989 | 145 | |
Z14978.1 | - | 8 - 9 | 971 | 145 |