The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006153.5 | NP_006144.1 | 1 - 2 | 115 | 56 |
GenBank mRNA | AK312692.1 | - | 1 - 2 | 59 | 56 |
BC006403.2 | - | 1 - 2 | 92 | 56 | |
DA265476.1 | - | 1 - 2 | 115 | 56 | |
DB117533.1 | - | 1 - 2 | 115 | 56 | |
X17576.1 | - | 1 - 2 | 86 | 56 |