The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022361.4 | NP_071756.2 | 3 - 4 | 990 | 121 |
NR_024539.1 | 3 - 4 | 531 | 121 | ||
XM_011536067.2 | XP_011534369.1 | 3 - 4 | 1150 | 121 | |
XM_017011194.1 | XP_016866683.1 | 3 - 4 | 2433 | 121 | |
XM_017011195.1 | XP_016866684.1 | 3 - 4 | 1893 | 121 | |
GenBank mRNA | AK225989.1 | - | 3 - 4 | 531 | 121 |
AK314101.1 | - | 3 - 4 | 990 | 121 | |
BC022323.2 | - | 2 - 3 | 840 | 121 |