Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
METTL21A
Entrez Gene ID
151194
Gene Name
methyltransferase like 21A
Gene Aliases
FAM119A, HCA557b, HSPA-KMT
Chromosome Location
Chr.2: 207580631 - 207625928 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001127395.2 NP_001120867.1 3 - 4 555 140
NM_145280.5 NP_660323.3 3 - 4 416 140
XM_005246339.3 XP_005246396.1 3 - 4 611 140
XM_005246340.3 XP_005246397.1 3 - 4 430 140
XM_005246341.2 XP_005246398.1 3 - 4 537 140
XM_006712329.3 XP_006712392.1 3 - 4 445 140
XM_011510725.2 XP_011509027.1 3 - 4 394 140
GenBank mRNA AF455817.1 - 3 - 4 367 140
AK093812.1 - 3 - 4 416 140
BC033720.1 - 3 - 4 427 140
BG832122.1 - 3 - 4 473 140

View Full Product Details