The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000133.3 | NP_000124.1 | 4 - 5 | 412 | 109 |
GenBank mRNA | A13997.1 | - | 4 - 5 | 384 | 109 |
AK292749.1 | - | 4 - 5 | 383 | 109 | |
BC109214.1 | - | 4 - 5 | 412 | 109 | |
BC109215.1 | - | 4 - 5 | 412 | 109 | |
FR846238.1 | - | 4 - 5 | 383 | 109 | |
FR846239.1 | - | 4 - 5 | 383 | 109 | |
J00136.1 | - | 4 - 5 | 383 | 109 | |
J00137.1 | - | 4 - 5 | 384 | 109 | |
M11309.1 | - | 4 - 5 | 383 | 109 | |
M35672.1 | - | 3 - 4 | 278 | 109 |