The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_194313.2 | NP_919289.2 | 12 - 13 | 4083 | 135 |
XM_005251451.2 | XP_005251508.1 | 8 - 9 | 2976 | 135 | |
XM_006716764.2 | XP_006716827.1 | 6 - 7 | 2687 | 135 | |
XM_011517861.1 | XP_011516163.1 | 13 - 14 | 4304 | 135 | |
XM_011517863.2 | XP_011516165.1 | 12 - 13 | 5393 | 135 | |
XM_017014697.1 | XP_016870186.1 | 12 - 13 | 4397 | 135 | |
XM_017014698.1 | XP_016870187.1 | 11 - 12 | 4170 | 135 | |
GenBank mRNA | AK125180.1 | - | 2 - 3 | 797 | 135 |
AK125872.1 | - | 3 - 4 | 2459 | 135 | |
BC048311.1 | - | 3 - 4 | 830 | 135 | |
BC110502.1 | - | 3 - 4 | 2441 | 135 | |
BC110503.1 | - | 3 - 4 | 2441 | 135 |