The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001177676.1 | NP_001171147.1 | 2 - 2 | 2070 | 123 |
NM_003485.3 | NP_003476.3 | 2 - 2 | 2024 | 123 | |
XM_005268110.4 | XP_005268167.1 | 2 - 2 | 3210 | 123 | |
XM_005268111.3 | XP_005268168.1 | 2 - 2 | 1990 | 123 | |
XM_005268112.3 | XP_005268169.1 | 2 - 2 | 1930 | 123 | |
XM_006720262.3 | XP_006720325.1 | 2 - 2 | 1956 | 123 | |
XM_011537196.2 | XP_011535498.1 | 3 - 3 | 2074 | 123 | |
XM_011537197.2 | XP_011535499.1 | 2 - 2 | 2354 | 123 | |
XM_011537198.2 | XP_011535500.1 | 2 - 2 | 2580 | 123 | |
XM_011537199.2 | XP_011535501.1 | 2 - 2 | 2025 | 123 | |
GenBank mRNA | BC098567.1 | - | 2 - 2 | 1967 | 123 |