The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001161368.1 | NP_001154840.1 | 9 - 10 | 868 | 158 |
NM_174977.3 | NP_777637.1 | 9 - 10 | 868 | 158 | |
XM_006724235.2 | XP_006724298.1 | 9 - 10 | 884 | 158 | |
XM_011530164.2 | XP_011528466.1 | 9 - 10 | 1048 | 158 | |
XM_017028787.1 | XP_016884276.1 | 8 - 9 | 997 | 158 | |
XM_017028789.1 | XP_016884278.1 | 8 - 9 | 670 | 158 | |
XM_017028791.1 | XP_016884280.1 | 8 - 9 | 697 | 158 | |
GenBank mRNA | AK092002.1 | - | 10 - 11 | 917 | 158 |
AK093418.1 | - | 9 - 10 | 885 | 158 | |
AK093448.1 | - | 9 - 10 | 852 | 158 | |
AY158085.1 | - | 9 - 10 | 768 | 158 | |
BC136358.1 | - | 9 - 10 | 840 | 158 | |
BC136359.1 | - | 9 - 10 | 840 | 158 | |
BC139912.1 | - | 9 - 10 | 807 | 158 | |
CR456346.1 | - | 7 - 8 | 627 | 158 |