The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_011530164.2 | XP_011528466.1 | 2 - 3 | 402 | 130 |
GenBank mRNA | AK092002.1 | - | 3 - 4 | 271 | 130 |