Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TRIM72
Entrez Gene ID
493829
Gene Name
tripartite motif containing 72
Gene Aliases
MG53
Chromosome Location
Chr.16: 31214021 - 31225656 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008274.3 NP_001008275.2 3 - 4 769 105
XM_017023254.1 XP_016878743.1 2 - 3 305 105
GenBank mRNA AK090695.1 - 2 - 3 214 105
AK131485.1 - 3 - 4 714 105
BC033211.1 - 3 - 4 699 105
DA058421.1 - 2 - 3 214 105

View Full Product Details