Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CSMD2
Entrez Gene ID
114784
Gene Name
CUB and Sushi multiple domains 2
Gene Aliases
dJ1007G16.1, dJ1007G16.2, dJ947L8.1
Chromosome Location
Chr.1: 33513999 - 34165842 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001281956.1 NP_001268885.1 33 - 34 5478 95
NM_052896.4 NP_443128.2 33 - 34 5210 95
XM_017000185.1 XP_016855674.1 33 - 34 5472 95
XM_017000186.1 XP_016855675.1 33 - 34 5472 95
XM_017000187.1 XP_016855676.1 33 - 34 5301 95
XM_017000188.1 XP_016855677.1 33 - 34 5210 95
XM_017000189.1 XP_016855678.1 33 - 34 5472 95
XM_017000190.1 XP_016855679.1 27 - 28 4737 95
XM_017000191.1 XP_016855680.1 33 - 34 5472 95
XM_017000192.1 XP_016855681.1 33 - 34 5472 95
XM_017000193.1 XP_016855682.1 33 - 34 5472 95
XM_017000194.1 XP_016855683.1 33 - 34 5210 95
GenBank mRNA AB212622.1 - 33 - 34 5478 95
AK122603.1 - 1 - 2 104 95
AK127722.1 - 12 - 13 2142 95
AK291019.1 - 12 - 13 1983 95
AY210418.1 - 33 - 34 5210 95

View Full Product Details