The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001281956.1 | NP_001268885.1 | 40 - 41 | 6314 | 113 |
NM_052896.4 | NP_443128.2 | 40 - 41 | 6046 | 113 | |
XM_017000185.1 | XP_016855674.1 | 40 - 41 | 6308 | 113 | |
XM_017000186.1 | XP_016855675.1 | 40 - 41 | 6308 | 113 | |
XM_017000187.1 | XP_016855676.1 | 40 - 41 | 6137 | 113 | |
XM_017000188.1 | XP_016855677.1 | 40 - 41 | 6046 | 113 | |
XM_017000189.1 | XP_016855678.1 | 40 - 41 | 6308 | 113 | |
XM_017000190.1 | XP_016855679.1 | 34 - 35 | 5573 | 113 | |
XM_017000191.1 | XP_016855680.1 | 40 - 41 | 6308 | 113 | |
XM_017000192.1 | XP_016855681.1 | 40 - 41 | 6308 | 113 | |
XM_017000193.1 | XP_016855682.1 | 40 - 41 | 6308 | 113 | |
XM_017000194.1 | XP_016855683.1 | 40 - 41 | 6046 | 113 | |
GenBank mRNA | AB212622.1 | - | 40 - 41 | 6314 | 113 |
AK122603.1 | - | 8 - 9 | 940 | 113 | |
AK127722.1 | - | 19 - 20 | 2978 | 113 | |
AK291019.1 | - | 19 - 20 | 2819 | 113 | |
AY210418.1 | - | 40 - 41 | 6046 | 113 |