The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_031861.2 | NP_114067.1 | 1 - 1 | 3970 | 104 |
GenBank mRNA | AF152476.1 | - | 1 - 1 | 2379 | 104 |
BC136706.1 | - | 1 - 1 | 3300 | 104 |