The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001083536.1 | NP_001077005.1 | 12 - 13 | 2016 | 131 |
NM_001286993.1 | NP_001273922.1 | 12 - 13 | 1998 | 131 | |
NM_033086.2 | NP_149077.2 | 12 - 13 | 1916 | 131 | |
XM_017015274.1 | XP_016870763.1 | 12 - 13 | 1758 | 131 | |
GenBank mRNA | AK000004.1 | - | 11 - 12 | 1593 | 131 |
AK225843.1 | - | 12 - 13 | 1668 | 131 | |
AK292676.1 | - | 12 - 13 | 1998 | 131 | |
AK301974.1 | - | 4 - 5 | 372 | 131 | |
AY211386.1 | - | 12 - 13 | 1852 | 131 | |
BC032429.1 | - | 12 - 13 | 1735 | 131 | |
BC111054.1 | - | 12 - 13 | 1853 | 131 | |
BX647829.1 | - | 12 - 13 | 1871 | 131 |