Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FGD3
Entrez Gene ID
89846
Gene Name
FYVE, RhoGEF and PH domain containing 3
Gene Aliases
ZFYVE5
Chromosome Location
Chr.9: 92947319 - 93036236 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001083536.1 NP_001077005.1 13 - 14 2124 170
NM_001286993.1 NP_001273922.1 13 - 14 2106 170
NM_033086.2 NP_149077.2 13 - 14 2024 170
XM_017015274.1 XP_016870763.1 13 - 14 1866 170
GenBank mRNA AK000004.1 - 12 - 13 1701 170
AK225843.1 - 13 - 14 1776 170
AK292676.1 - 13 - 14 2106 170
AK301974.1 - 5 - 6 480 170
AY211386.1 - 13 - 14 1960 170
BC032429.1 - 13 - 14 1843 170
BC111054.1 - 13 - 14 1961 170
BX647829.1 - 13 - 14 1979 170

View Full Product Details