The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001083536.1 | NP_001077005.1 | 4 - 5 | 1174 | 95 |
NM_001286993.1 | NP_001273922.1 | 4 - 5 | 1156 | 95 | |
NM_033086.2 | NP_149077.2 | 4 - 5 | 1074 | 95 | |
XM_017015274.1 | XP_016870763.1 | 4 - 5 | 916 | 95 | |
GenBank mRNA | AK000004.1 | - | 3 - 4 | 751 | 95 |
AK225843.1 | - | 4 - 5 | 826 | 95 | |
AK292676.1 | - | 4 - 5 | 1156 | 95 | |
AY211386.1 | - | 4 - 5 | 1010 | 95 | |
BC032429.1 | - | 4 - 5 | 893 | 95 | |
BC111054.1 | - | 4 - 5 | 1011 | 95 | |
BX647829.1 | - | 4 - 5 | 1029 | 95 |