Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
STXBP6
Entrez Gene ID
29091
Gene Name
syntaxin binding protein 6
Gene Aliases
HSPC156, amisyn
Chromosome Location
Chr.14: 24809654 - 25050297 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK096957.1 - 4 - 5 458 124

View Full Product Details