Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- ASXL3
- Entrez Gene ID
-
80816
- Gene Name
- additional sex combs like 3, transcriptional regulator
- Gene Aliases
-
BRPS, KIAA1713
- Chromosome Location
-
Chr.18: 33578577 - 33751195 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_030632.2 |
NP_085135.1 |
2 - 3 |
193 |
139 |
View Full Product Details