Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RNF14
Entrez Gene ID
9604
Gene Name
ring finger protein 14
Gene Aliases
ARA54, HFB30, HRIHFB2038, TRIAD2
Chromosome Location
Chr.5: 141949337 - 141993709 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001201365.1 NP_001188294.1 4 - 5 554 77
NM_004290.4 NP_004281.1 4 - 5 768 77
NM_183399.2 NP_899646.1 3 - 4 380 77
NM_183400.2 NP_899647.1 3 - 4 594 77
NM_183401.2 NP_899648.1 4 - 5 574 77
XM_005268536.2 XP_005268593.1 4 - 5 871 77
XM_005268537.2 XP_005268594.1 4 - 5 915 77
XM_011537714.2 XP_011536016.1 3 - 4 385 77
XM_017010070.1 XP_016865559.1 4 - 5 519 77
GenBank mRNA AB022663.1 - 4 - 5 542 77
AF060544.1 - 3 - 4 345 77
AK023698.1 - 3 - 4 344 77
AK023884.1 - 4 - 5 510 77
BC126185.1 - 4 - 5 513 77
BC144061.1 - 4 - 5 513 77
BX647955.1 - 4 - 5 541 77
CB997918.1 - 3 - 4 402 77
CD106708.1 - 4 - 5 580 77
CR456702.1 - 2 - 3 306 77

View Full Product Details