The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001201365.1 | NP_001188294.1 | 5 - 6 | 1080 | 132 |
NM_004290.4 | NP_004281.1 | 5 - 6 | 1294 | 132 | |
NM_183398.2 | NP_899645.1 | 4 - 5 | 1142 | 132 | |
NM_183399.2 | NP_899646.1 | 4 - 5 | 906 | 132 | |
NM_183400.2 | NP_899647.1 | 4 - 5 | 1120 | 132 | |
NM_183401.2 | NP_899648.1 | 5 - 6 | 1100 | 132 | |
XM_005268536.2 | XP_005268593.1 | 5 - 6 | 1397 | 132 | |
XM_005268537.2 | XP_005268594.1 | 5 - 6 | 1441 | 132 | |
XM_005268539.2 | XP_005268596.1 | 4 - 5 | 910 | 132 | |
XM_005268540.3 | XP_005268597.1 | 4 - 5 | 1066 | 132 | |
XM_005268541.3 | XP_005268598.1 | 3 - 4 | 822 | 132 | |
XM_011537714.2 | XP_011536016.1 | 4 - 5 | 911 | 132 | |
XM_017010070.1 | XP_016865559.1 | 5 - 6 | 1045 | 132 | |
XM_017010071.1 | XP_016865560.1 | 4 - 5 | 1272 | 132 | |
XM_017010072.1 | XP_016865561.1 | 4 - 5 | 1232 | 132 | |
GenBank mRNA | AB022663.1 | - | 5 - 6 | 1068 | 132 |
AF060544.1 | - | 4 - 5 | 871 | 132 | |
AK023698.1 | - | 4 - 5 | 870 | 132 | |
AK023884.1 | - | 5 - 6 | 1036 | 132 | |
AK057868.1 | - | 4 - 5 | 918 | 132 | |
AK129515.1 | - | 4 - 5 | 1071 | 132 | |
BC126185.1 | - | 5 - 6 | 1039 | 132 | |
BC144061.1 | - | 5 - 6 | 1039 | 132 | |
BX647955.1 | - | 5 - 6 | 1067 | 132 | |
CR456702.1 | - | 3 - 4 | 832 | 132 |