Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RNF14
Entrez Gene ID
9604
Gene Name
ring finger protein 14
Gene Aliases
ARA54, HFB30, HRIHFB2038, TRIAD2
Chromosome Location
Chr.5: 141949337 - 141993709 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001201365.1 NP_001188294.1 6 - 7 1315 108
NM_004290.4 NP_004281.1 6 - 7 1529 108
NM_183398.2 NP_899645.1 5 - 6 1377 108
NM_183399.2 NP_899646.1 5 - 6 1141 108
NM_183400.2 NP_899647.1 5 - 6 1355 108
NM_183401.2 NP_899648.1 6 - 7 1335 108
XM_005268536.2 XP_005268593.1 6 - 7 1632 108
XM_005268537.2 XP_005268594.1 6 - 7 1676 108
XM_005268539.2 XP_005268596.1 5 - 6 1145 108
XM_005268540.3 XP_005268597.1 5 - 6 1301 108
XM_005268541.3 XP_005268598.1 4 - 5 1057 108
XM_005268542.2 XP_005268599.1 5 - 6 844 108
XM_005268543.4 XP_005268600.1 4 - 5 650 108
XM_011537714.2 XP_011536016.1 5 - 6 1146 108
XM_017010070.1 XP_016865559.1 6 - 7 1280 108
XM_017010071.1 XP_016865560.1 5 - 6 1507 108
XM_017010072.1 XP_016865561.1 5 - 6 1467 108
XM_017010073.1 XP_016865562.1 5 - 6 882 108
XM_017010074.1 XP_016865563.1 5 - 6 1148 108
XM_017010075.1 XP_016865564.1 5 - 6 807 108
XM_017010076.1 XP_016865565.1 5 - 6 793 108
GenBank mRNA AB015333.1 - 1 - 2 212 108
AB022663.1 - 6 - 7 1303 108
AF060544.1 - 5 - 6 1106 108
AK023698.1 - 5 - 6 1105 108
AK023884.1 - 6 - 7 1271 108
AK057868.1 - 5 - 6 1153 108
AK129515.1 - 5 - 6 1306 108
AK294262.1 - 5 - 6 746 108
BC126185.1 - 6 - 7 1274 108
BC144061.1 - 6 - 7 1274 108
BX647955.1 - 6 - 7 1302 108
CR456702.1 - 4 - 5 1067 108

View Full Product Details