The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002106.3 | NP_002097.1 | 1 - 2 | 194 | 145 |
GenBank mRNA | AK056803.1 | - | 1 - 2 | 194 | 145 |
BC103743.1 | - | 1 - 2 | 229 | 145 | |
CD685635.1 | - | 1 - 2 | 227 | 145 |