The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_207581.3 | NP_997464.2 | 5 - 6 | 1055 | 155 |
XM_017022180.1 | XP_016877669.1 | 5 - 6 | 1110 | 155 | |
GenBank mRNA | BC137465.1 | - | 5 - 6 | 976 | 155 |
BX537581.1 | - | 5 - 6 | 1339 | 155 | |
DQ489734.1 | - | 5 - 6 | 906 | 155 |