Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CYP2C18
Entrez Gene ID
1562
Gene Name
cytochrome P450 family 2 subfamily C member 18
Gene Aliases
CPCI, CYP2C, CYP2C17, P450-6B/29C, P450IIC17
Chromosome Location
Chr.10: 94683494 - 94736190 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000772.2 NP_000763.1 2 - 3 656 120
NM_001128925.1 NP_001122397.1 2 - 3 656 120
GenBank mRNA AK223510.1 - 2 - 3 355 120
AK308558.1 - 2 - 3 311 120
AK313403.1 - 2 - 3 355 120
BC069666.1 - 2 - 3 524 120
BC096257.1 - 2 - 3 375 120
BC096258.1 - 2 - 3 375 120
BC096260.1 - 2 - 3 375 120
M61853.1 - 2 - 3 372 120
M61856.1 - 2 - 3 529 120

View Full Product Details