The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017590.5 | NP_060060.3 | 15 - 16 | 2047 | 156 |
XM_011530016.2 | XP_011528318.1 | 15 - 16 | 1969 | 156 | |
GenBank mRNA | AB028954.1 | - | 12 - 13 | 1660 | 156 |
AF188530.1 | - | 15 - 16 | 1875 | 156 | |
AK315009.1 | - | 15 - 16 | 1989 | 156 | |
BC024313.2 | - | 6 - 7 | 714 | 156 | |
BC098378.1 | - | 15 - 16 | 1985 | 156 | |
BC152558.1 | - | 15 - 16 | 2025 | 156 |