The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017590.5 | NP_060060.3 | 18 - 19 | 2447 | 150 |
XM_011530016.2 | XP_011528318.1 | 18 - 19 | 2369 | 150 | |
GenBank mRNA | AB028954.1 | - | 15 - 16 | 2060 | 150 |
AK315009.1 | - | 18 - 19 | 2389 | 150 | |
BC024313.2 | - | 9 - 10 | 1114 | 150 | |
BC098378.1 | - | 18 - 19 | 2385 | 150 | |
BC152558.1 | - | 18 - 19 | 2425 | 150 |