The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017590.5 | NP_060060.3 | 4 - 5 | 558 | 116 |
XM_011530016.2 | XP_011528318.1 | 4 - 5 | 539 | 116 | |
GenBank mRNA | AB028954.1 | - | 1 - 2 | 171 | 116 |
AK023849.1 | - | 4 - 5 | 346 | 116 | |
AK225891.1 | - | 4 - 5 | 531 | 116 | |
AK315009.1 | - | 4 - 5 | 500 | 116 | |
BC098378.1 | - | 4 - 5 | 496 | 116 | |
BC152558.1 | - | 4 - 5 | 536 | 116 |