The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017590.5 | NP_060060.3 | 5 - 6 | 719 | 152 |
XM_011530016.2 | XP_011528318.1 | 5 - 6 | 700 | 152 | |
GenBank mRNA | AB028954.1 | - | 2 - 3 | 332 | 152 |
AK023849.1 | - | 5 - 6 | 507 | 152 | |
AK225891.1 | - | 5 - 6 | 692 | 152 | |
AK315009.1 | - | 5 - 6 | 661 | 152 | |
BC098378.1 | - | 5 - 6 | 657 | 152 | |
BC152558.1 | - | 5 - 6 | 697 | 152 |