Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TRIM67
Entrez Gene ID
440730
Gene Name
tripartite motif containing 67
Gene Aliases
TNL
Chromosome Location
Chr.1: 231162928 - 231221568 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001004342.3 NP_001004342.3 4 - 5 1412 158
NM_001300889.1 NP_001287818.1 6 - 7 1226 158
XM_011544192.2 XP_011542494.1 4 - 5 1412 158
XM_017001323.1 XP_016856812.1 4 - 5 1412 158
GenBank mRNA AK126782.1 - 4 - 5 1412 158
AY253917.1 - 6 - 7 1184 158

View Full Product Details