Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
OLA1
Entrez Gene ID
29789
Gene Name
Obg-like ATPase 1
Gene Aliases
DOC45, GBP45, GTBP9, GTPBP9, PTD004
Chromosome Location
Chr.2: 174072447 - 174248637 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AF078859.1 - 8 - 9 990 156
AF078868.1 - 8 - 9 971 156
AF116703.2 - 5 - 6 548 156
AF134478.1 - 8 - 9 946 156
AK027523.1 - 8 - 9 1023 156
AK074710.1 - 7 - 8 921 156
AK225112.1 - 8 - 9 896 156
AK225138.1 - 8 - 9 999 156
AL136546.1 - 7 - 8 878 156
AL833441.1 - 8 - 9 987 156
BC012842.1 - 8 - 9 1060 156
BC029376.1 - 8 - 9 939 156
BC091522.1 - 8 - 9 914 156
DQ250006.1 - 8 - 9 915 156

View Full Product Details