The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016038.2 | NP_057122.2 | 4 - 5 | 810 | 130 |
GenBank mRNA | AF151855.1 | - | 4 - 5 | 796 | 130 |
AK001779.1 | - | 4 - 5 | 810 | 130 | |
AK289609.1 | - | 4 - 5 | 815 | 130 | |
AY169963.1 | - | 4 - 5 | 810 | 130 | |
BC065700.1 | - | 4 - 5 | 780 | 130 | |
KU178629.1 | - | 4 - 5 | 626 | 130 | |
KU178630.1 | - | 3 - 4 | 496 | 130 |