The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001006641.3 | NP_001006642.1 | 6 - 7 | 1020 | 113 |
NM_001006642.3 | NP_001006643.1 | 6 - 7 | 1265 | 113 | |
NM_001265614.2 | NP_001252543.1 | 7 - 8 | 1301 | 113 | |
NM_052901.4 | NP_443133.2 | 6 - 7 | 883 | 113 | |
NR_049766.2 | 6 - 7 | 926 | 113 | ||
XM_005251688.1 | XP_005251745.1 | 7 - 8 | 1056 | 113 | |
XM_005251689.4 | XP_005251746.1 | 6 - 7 | 1279 | 113 | |
XM_006716948.2 | XP_006717011.1 | 7 - 8 | 1457 | 113 | |
XM_006716949.3 | XP_006717012.1 | 7 - 8 | 1317 | 113 | |
GenBank mRNA | AB067483.1 | - | 7 - 8 | 1300 | 113 |
AJ619963.1 | - | 6 - 7 | 1184 | 113 | |
AJ619989.1 | - | 6 - 7 | 808 | 113 | |
AJ619990.1 | - | 6 - 7 | 903 | 113 | |
AJ619991.1 | - | 6 - 7 | 865 | 113 | |
AJ619992.1 | - | 6 - 7 | 851 | 113 | |
AK290705.1 | - | 6 - 7 | 856 | 113 | |
AK290991.1 | - | 6 - 7 | 1020 | 113 | |
AY358515.1 | - | 6 - 7 | 858 | 113 | |
BC005163.2 | - | 2 - 3 | 317 | 113 | |
BC089448.1 | - | 6 - 7 | 874 | 113 | |
BC103930.1 | - | 6 - 7 | 825 | 113 | |
BC103931.1 | - | 6 - 7 | 825 | 113 | |
BC103932.1 | - | 6 - 7 | 825 | 113 | |
BC103933.1 | - | 6 - 7 | 825 | 113 |