Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KRT37
Entrez Gene ID
8688
Gene Name
keratin 37
Gene Aliases
HA7, K37, KRTHA7
Chromosome Location
Chr.17: 41420557 - 41424570 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003770.4 NP_003761.3 6 - 7 1288 179
GenBank mRNA AJ786655.1 - 6 - 7 1124 179

View Full Product Details