The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001284358.1 | NP_001271287.1 | 5 - 6 | 773 | 153 |
NM_001284362.1 | NP_001271291.1 | 7 - 8 | 956 | 153 | |
NM_033030.5 | NP_149019.1 | 7 - 8 | 869 | 153 | |
NM_197970.2 | NP_932074.1 | 7 - 8 | 806 | 153 | |
XM_006712715.3 | XP_006712778.1 | 7 - 8 | 1166 | 153 | |
GenBank mRNA | AF272858.1 | - | 7 - 8 | 814 | 153 |
AK058099.1 | - | 7 - 8 | 869 | 153 | |
AK097795.1 | - | 7 - 8 | 1165 | 153 | |
AK098824.1 | - | 7 - 8 | 838 | 153 | |
AK302817.1 | - | 5 - 6 | 773 | 153 | |
AM295005.1 | - | 7 - 8 | 956 | 153 | |
BC033674.1 | - | 7 - 8 | 821 | 153 |