Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CCDC178
Entrez Gene ID
374864
Gene Name
coiled-coil domain containing 178
Gene Aliases
C18orf34
Chromosome Location
Chr.18: 32937402 - 33441040 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105528.1 NP_001098998.1 10 - 11 1157 174
NM_001308126.1 NP_001295055.1 10 - 11 1157 174
NM_198995.2 NP_945346.2 11 - 12 1226 174
XM_011525948.1 XP_011524250.1 11 - 12 1226 174
XM_011525951.1 XP_011524253.1 11 - 12 1226 174
XM_011525954.2 XP_011524256.1 11 - 12 1226 174
XM_011525955.1 XP_011524257.1 11 - 12 1226 174
XM_017025721.1 XP_016881210.1 11 - 12 1226 174
XM_017025722.1 XP_016881211.1 11 - 12 1247 174
XM_017025723.1 XP_016881212.1 11 - 12 1256 174
XM_017025724.1 XP_016881213.1 5 - 6 790 174
XM_017025725.1 XP_016881214.1 5 - 6 1407 174
GenBank mRNA BC091517.1 - 11 - 12 1198 174
BC130535.1 - 10 - 11 1094 174
BC144178.1 - 10 - 11 1094 174

View Full Product Details