Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CCDC178
Entrez Gene ID
374864
Gene Name
coiled-coil domain containing 178
Gene Aliases
C18orf34
Chromosome Location
Chr.18: 32937402 - 33441040 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105528.1 NP_001098998.1 14 - 15 1730 153
NM_001308126.1 NP_001295055.1 14 - 15 1730 153
NM_198995.2 NP_945346.2 15 - 16 1799 153
XM_011525948.1 XP_011524250.1 15 - 16 1799 153
XM_011525951.1 XP_011524253.1 15 - 16 1799 153
XM_017025721.1 XP_016881210.1 15 - 16 1799 153
XM_017025722.1 XP_016881211.1 15 - 16 1820 153
XM_017025723.1 XP_016881212.1 15 - 16 1829 153
XM_017025724.1 XP_016881213.1 9 - 10 1363 153
XM_017025725.1 XP_016881214.1 9 - 10 1980 153
GenBank mRNA AK126038.1 - 15 - 16 1799 153
AK129955.1 - 2 - 3 229 153
BC091517.1 - 15 - 16 1771 153
BC130535.1 - 14 - 15 1667 153
BC144178.1 - 14 - 15 1667 153

View Full Product Details