Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EML5
Entrez Gene ID
161436
Gene Name
echinoderm microtubule associated protein like 5
Gene Aliases
EMAP-2
Chromosome Location
Chr.14: 88614213 - 88792752 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_183387.2 NP_899243.1 18 - 19 2995 132
XM_006720070.3 XP_006720133.1 18 - 19 2795 132
XM_011536528.2 XP_011534830.1 18 - 19 2796 132
XM_011536530.2 XP_011534832.1 18 - 19 2795 132
XM_011536531.2 XP_011534833.1 18 - 19 2795 132
XM_011536532.2 XP_011534834.1 18 - 19 2795 132
XM_011536533.2 XP_011534835.1 18 - 19 2681 132
XM_011536534.2 XP_011534836.1 19 - 20 2713 132
XM_011536536.2 XP_011534838.1 18 - 19 2795 132
XM_011536538.2 XP_011534840.1 18 - 19 2795 132
XM_017021062.1 XP_016876551.1 18 - 19 2795 132
XM_017021063.1 XP_016876552.1 18 - 19 2795 132
XM_017021064.1 XP_016876553.1 18 - 19 2681 132
XM_017021065.1 XP_016876554.1 18 - 19 2795 132
XM_017021066.1 XP_016876555.1 17 - 18 2596 132
XM_017021067.1 XP_016876556.1 18 - 19 3116 132
XM_017021068.1 XP_016876557.1 18 - 19 2795 132
XM_017021069.1 XP_016876558.1 18 - 19 3040 132
XM_017021070.1 XP_016876559.1 18 - 19 2795 132
GenBank mRNA AK128086.1 - 11 - 12 1534 132
AY357725.1 - 9 - 10 1245 132
BC150639.1 - 18 - 19 2995 132

View Full Product Details