Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- EML5
- Entrez Gene ID
-
161436
- Gene Name
- echinoderm microtubule associated protein like 5
- Gene Aliases
-
EMAP-2
- Chromosome Location
-
Chr.14: 88614213 - 88792752 on Build GRCh38
View Full Product Details